Anti-LYVE1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Novus Biologicals
NB100-726H
NOVUNB100-726HEA 530 EUR
NOVUNB100-726H
Anti-LYVE1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Primary
Antigen: LYVE1 (lymphatic vessel endothelial hyaluronan receptor 1)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse