Anti-NK1.1 Mouse Monoclonal Antibody

Lieferant: Novus Biologicals
NB100-77528
NOVUNB100-77528EA 548 EUR
NOVUNB100-77528
Anti-NK1.1 Mouse Monoclonal Antibody
Antikörper
Mouse Monoclonal NK1.1 Antibody (PK136). Tested Applications: Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoprecipitation. Tested Reactivity: Mouse.

Type: Primary
Antigen: NK1.1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR