Anti-RANBP2 Rabbit polyclonal antibody

Lieferant: Novus Biologicals
NB100-93338
NOVUNB100-93338EA 684 EUR
NOVUNB100-93338
Anti-RANBP2 Rabbit polyclonal antibody
Antikörper
The RanBP2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to RanBP2. This antibody reacts with human. The RanBP2 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: RanBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR