Anti-C5 Mouse Monoclonal Antibody

Lieferant: Novus Biologicals
NB110-12179
NOVUNB110-12179EA 551 EUR
NOVUNB110-12179
Anti-C5 Mouse Monoclonal Antibody
Antikörper
Mouse Monoclonal Complement C5 Antibody (557). Tested Applications: Western Blot, Blocking/Neutralizing, ELISA, Functional. Tested Reactivity: Human.

Type: Primary
Antigen: C5
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR