Anti-DISC1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Novus Biologicals
NB110-40773B
NOVUNB110-40773BEA 668 EUR
NOVUNB110-40773B
Anti-DISC1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: DISC1 (disrupted in schizophrenia 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR