Anti-GPIHBP1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Lieferant: Novus Biologicals
NB110-41539F
NOVUNB110-41539FEA 722 EUR
NOVUNB110-41539F
Anti-GPIHBP1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: GPIHBP1 (glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse