Anti-SAT1 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB110-41622 NB110-41622SS
NOVUNB110-41622EA 647 EUR
NOVUNB110-41622 NOVUNB110-41622SS
Anti-SAT1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SAT1 (spermidine/spermine N1-acetyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR