Anti-ATG5 Rabbit Polyclonal Antibody (DyLight® 488)

Lieferant: Novus Biologicals
NB110-53818G
NOVUNB110-53818GEA 768 EUR
NOVUNB110-53818G
Anti-ATG5 Rabbit Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Primary
Antigen: ATG5 (autophagy related 5)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR