Anti-ROBO4 Rabbit Polyclonal Antibody (DyLight 488)

Lieferant: Novus Biologicals
NB110-58778G
NOVUNB110-58778GEA 668 EUR
NOVUNB110-58778G
Anti-ROBO4 Rabbit Polyclonal Antibody (DyLight 488)
Antikörper
The ROBO4 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to ROBO4. This antibody reacts with human. The ROBO4 Antibody [DyLight 488] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: ROBO4
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR