Anti-KIAA1524 Mouse Monoclonal Antibody (DyLight® 755) [clone: 2G10]

Lieferant: Novus Biologicals
NB110-59722IR
NOVUNB110-59722IREA 722 EUR
NOVUNB110-59722IR
Anti-KIAA1524 Mouse Monoclonal Antibody (DyLight® 755) [clone: 2G10]
Antikörper
Type: Primary
Antigen: KIAA1524 (KIAA1524)
Clonality: monoclonal
Clone: 2G10
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR