Anti-CD36 Mouse Monoclonal Antibody (DyLight® 488) [clone: D-2712]
Lieferant: Novus Biologicals
NB110-59724G
NOVUNB110-59724GEA 731 EUR
NOVUNB110-59724G
Anti-CD36 Mouse Monoclonal Antibody (DyLight® 488) [clone: D-2712]
Antikörper
Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: monoclonal
Clone: D-2712
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgA Kappa
Reactivity: Mouse, Rat