Anti-Capicua Rabbit Polyclonal Antibody (DyLight® 650)

Lieferant: Novus Biologicals
NB110-59905C
NOVUNB110-59905CEA 668 EUR
NOVUNB110-59905C
Anti-Capicua Rabbit Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Primary
Antigen: Capicua
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR