Anti-Capicua Rabbit Polyclonal Antibody (Biotin)

Lieferant: Novus Biologicals
NB110-59906B
NOVUNB110-59906BEA 803 EUR
NOVUNB110-59906B
Anti-Capicua Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Capicua
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR