Anti-WT1 Mouse Monoclonal Antibody (DyLight® 405) [clone: 6F-H2]
Lieferant: Novus Biologicals
NB110-60011V
NOVUNB110-60011VEA 679 EUR
NOVUNB110-60011V
Anti-WT1 Mouse Monoclonal Antibody (DyLight® 405) [clone: 6F-H2]
Antikörper
Type: Primary
Antigen: WT1 (Wilms tumor 1)
Clonality: monoclonal
Clone: 6F-H2
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human