Anti-PLIN5 Rabbit Polyclonal Antibody (Biotin)
Lieferant: Novus Biologicals
NB110-60511B
NOVUNB110-60511BEA 722 EUR
NOVUNB110-60511B
Anti-PLIN5 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: PLIN5 (perilipin 5)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse