Anti-LPO Rabbit Polyclonal Antibody (Biotin)

Lieferant: Novus Biologicals
NB110-61572B
NOVUNB110-61572BEA 722 EUR
NOVUNB110-61572B
Anti-LPO Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: LPO (lactoperoxidase)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR