Anti-HSPA13 Mouse Monoclonal Antibody ATTO 488 [clone: C92F3A-5]
Lieferant: Novus Biologicals
NB110-61582
NOVUNB110-61582EA 379 EUR
NOVUNB110-61582
Anti-HSPA13 Mouse Monoclonal Antibody ATTO 488 [clone: C92F3A-5]
Antikörper
Type: Primary
Antigen: HSPA13 (heat shock protein 70kDa family, member 13)
Clonality: monoclonal
Clone: C92F3A-5
Conjugation: ATTO 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen