Anti-NPM1 Rabbit Polyclonal Antibody (Biotin)
Lieferant: Novus Biologicals
NB110-61646B
NOVUNB110-61646BEA 723 EUR
NOVUNB110-61646B
Anti-NPM1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
The NPM1 Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to NPM1. This antibody reacts with human. The NPM1 Antibody [Biotin] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:50 - 1:100
Type: Primary
Antigen: NPM1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human