Anti-MAT2A Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: Novus Biologicals
NB110-94158F
NOVUNB110-94158FEA 722 EUR
NOVUNB110-94158F
Anti-MAT2A Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: MAT2A (methionine adenosyltransferase II, alpha)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR