Anti-Prostein Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB200-129
NOVUNB200-129EA 608 EUR
NOVUNB200-129
Anti-Prostein Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal Prostein Antibody. Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Type: Primary
Antigen: Prostein
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR