Anti-RNA Polymerase II Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 4H8]

Lieferant: Novus Biologicals
NB200-598H
NOVUNB200-598HEA 743 EUR
NOVUNB200-598H
Anti-RNA Polymerase II Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 4H8]
Antikörper
Type: Primary
Antigen: RNA polymerase II
Clonality: monoclonal
Clone: 4H8
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR