Anti-HIST3H3 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Novus Biologicals
NB21-1012B
NOVUNB21-1012BEA 836 EUR
NOVUNB21-1012B
Anti-HIST3H3 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR