Anti-HIST3H3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Lieferant: Novus Biologicals
NB21-1052F
NOVUNB21-1052FEA 836 EUR
NOVUNB21-1052F
Anti-HIST3H3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen