Anti-HIST3H3 Rabbit Polyclonal Antibody (DyLight® 488)
Lieferant: Novus Biologicals
NB21-1052G
NOVUNB21-1052GEA 836 EUR
NOVUNB21-1052G
Anti-HIST3H3 Rabbit Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen