Anti-Laminin Rabbit Polyclonal Antibody (DyLight® 550)
Lieferant: Novus Biologicals
NB300-144R
NOVUNB300-144REA 775 EUR
NOVUNB300-144R
Anti-Laminin Rabbit Polyclonal Antibody (DyLight® 550)
Antikörper
Type: Primary
Antigen: Laminin
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen