Anti-FXR2 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 1G2]

Lieferant: Novus Biologicals
NB300-259F
NOVUNB300-259FEA 655 EUR
NOVUNB300-259F
Anti-FXR2 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 1G2]
Antikörper
Type: Primary
Antigen: FXR2 (fragile X mental retardation, autosomal homolog 2)
Clonality: monoclonal
Clone: 1G2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR