Anti-NES Rabbit Polyclonal Antibody (Biotin)

Lieferant: Novus Biologicals
NB300-265B
NOVUNB300-265BEA 710 EUR
NOVUNB300-265B
Anti-NES Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: NES (nestin)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR