Anti-SLC7A11 Rabbit Polyclonal Antibody (DyLight® 405)

Lieferant: Novus Biologicals
NB300-317V
NOVUNB300-317VEA 706 EUR
NOVUNB300-317V
Anti-SLC7A11 Rabbit Polyclonal Antibody (DyLight® 405)
Antikörper
Type: Primary
Antigen: SLC7A11 (solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR