Anti-SLC7A11 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Novus Biologicals
NB300-318H
NOVUNB300-318HEA 827 EUR
NOVUNB300-318H
Anti-SLC7A11 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Primary
Antigen: SLC7A11 (solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR