Anti-eNOS Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB300-500
NOVUNB300-500EA 671 EUR
NOVUNB300-500
Anti-eNOS Rabbit Polyclonal Antibody
Antikörper
The eNOS Antibody from Novus Biologicals is a rabbit polyclonal antibody to eNOS. This antibody reacts with human, mouse, rat, porcine, amphibian, bovine, canine, drosophila. The eNOS Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: eNOS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: SLQERQLRGAVPWAFD
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Drosophila
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR