Anti-SCARB1 Rabbit Polyclonal Antibody (Biotin)
Lieferant: Novus Biologicals
NB400-104B
NOVUNB400-104BEA 751 EUR
NOVUNB400-104B
Anti-SCARB1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: SCARB1 (scavenger receptor class B, member 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen