Anti-ABCG8 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Novus Biologicals
NB400-110B
NOVUNB400-110BEA 591 EUR
NOVUNB400-110B
Anti-ABCG8 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: ABCG8 (ATP-binding cassette, sub-family G (WHITE), member 8)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR