Anti-Endothelial Lipase Rabbit Polyclonal Antibody (DyLight® 650)

Lieferant: Novus Biologicals
NB400-111C
NOVUNB400-111CEA 655 EUR
NOVUNB400-111C
Anti-Endothelial Lipase Rabbit Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Primary
Antigen: Endothelial Lipase
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR