Anti-ABCE1 Rabbit Polyclonal Antibody (DyLight® 405)
Lieferant: Novus Biologicals
NB400-116V
NOVUNB400-116VEA 710 EUR
NOVUNB400-116V
Anti-ABCE1 Rabbit Polyclonal Antibody (DyLight® 405)
Antikörper
Type: Primary
Antigen: ABCE1 (ATP-binding cassette, sub-family E (OABP), member 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human