Anti-Endothelial Lipase Rabbit Polyclonal Antibody (DyLight® 405)
Lieferant: Novus Biologicals
NB400-118V
NOVUNB400-118VEA 723 EUR
NOVUNB400-118V
Anti-Endothelial Lipase Rabbit Polyclonal Antibody (DyLight® 405)
Antikörper
Type: Primary
Antigen: Endothelial Lipase
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human