Anti-MRP1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: IU2H10]
Lieferant: Novus Biologicals
NB400-156F
NOVUNB400-156FEA 679 EUR
NOVUNB400-156F
Anti-MRP1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: IU2H10]
Antikörper
Type: Primary
Antigen: MRP1
Clonality: monoclonal
Clone: IU2H10
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat