Anti-ABCA7 Rat Monoclonal Antibody [clone: 7A1-144]

Lieferant: Novus Biologicals
NB400-163 NB400-163SS
NOVUNB400-163EA 591 EUR
NOVUNB400-163 NOVUNB400-163SS
Anti-ABCA7 Rat Monoclonal Antibody [clone: 7A1-144]
Antikörper
Type: Primary
Antigen: ABCA7 (ATP-binding cassette, sub-family A (ABC1), member 7)
Clonality: monoclonal
Clone: 7A1-144
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR