Anti-ABCA7 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 7A1-144]
Lieferant: Novus Biologicals
NB400-163F
NOVUNB400-163FEA 710 EUR
NOVUNB400-163F
Anti-ABCA7 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 7A1-144]
Antikörper
Type: Primary
Antigen: ABCA7 (ATP-binding cassette, sub-family A (ABC1), member 7)
Clonality: monoclonal
Clone: 7A1-144
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse