Anti-ABCA7 Rat Monoclonal Antibody (DyLight® 755) [clone: 7A1-144]
Lieferant: Novus Biologicals
NB400-163IR
NOVUNB400-163IREA 710 EUR
NOVUNB400-163IR
Anti-ABCA7 Rat Monoclonal Antibody (DyLight® 755) [clone: 7A1-144]
Antikörper
Type: Primary
Antigen: ABCA7 (ATP-binding cassette, sub-family A (ABC1), member 7)
Clonality: monoclonal
Clone: 7A1-144
Conjugation: DyLight® 755
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse