Anti-MAPK Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB500-138
NOVUNB500-138EA 632 EUR
NOVUNB500-138
Anti-MAPK Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal p38 [p Tyr182] Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Type: Primary
Antigen: MAPK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR