Anti-POT1 Rabbit Polyclonal Antibody (DyLight® 405)

Lieferant: Novus Biologicals
NB500-176V
NOVUNB500-176VEA 710 EUR
NOVUNB500-176V
Anti-POT1 Rabbit Polyclonal Antibody (DyLight® 405)
Antikörper
Type: Primary
Antigen: POT1 (protection of telomeres 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR