Anti-CD34 Rat Monoclonal Antibody (DyLight® 350) [clone: MEC 14.7]
Lieferant: Novus Biologicals
NB600-1071UV
NOVUNB600-1071UVEA 754 EUR
NOVUNB600-1071UV
Anti-CD34 Rat Monoclonal Antibody (DyLight® 350) [clone: MEC 14.7]
Antikörper
Type: Primary
Antigen: CD34 (CD34 molecule)
Clonality: monoclonal
Clone: MEC 14.7
Conjugation: DyLight® 350
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse