Anti-Ctip1 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NB600-261
NOVUNB600-261EA 730 EUR
NOVUNB600-261
Anti-Ctip1 Rabbit Polyclonal Antibody
Antikörper
The Ctip1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Ctip1. This antibody reacts with human, mouse. The Ctip1 Antibody has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunoprecipitation.

There is 1 scientific publication usiµg this antibody.

Type: Primary
Antigen: Ctip1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR