Anti-Aflatoxin B1 Mouse Monoclonal Antibody (DyLight® 755) [clone: 6A10]
Lieferant: Novus Biologicals
NB600-443IR
NOVUNB600-443IREA 710 EUR
NOVUNB600-443IR
Anti-Aflatoxin B1 Mouse Monoclonal Antibody (DyLight® 755) [clone: 6A10]
Antikörper
Type: Primary
Antigen: Aflatoxin B1 (aflatoxin B1)
Clonality: monoclonal
Clone: 6A10
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Pan