Anti-Aflatoxin B1 Mouse Monoclonal Antibody (DyLight® 405) [clone: 6A10]

Lieferant: Novus Biologicals
NB600-443V
NOVUNB600-443VEA 710 EUR
NOVUNB600-443V
Anti-Aflatoxin B1 Mouse Monoclonal Antibody (DyLight® 405) [clone: 6A10]
Antikörper
Type: Primary
Antigen: Aflatoxin B1 (aflatoxin B1)
Clonality: monoclonal
Clone: 6A10
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR