Anti-Ethenocytidine Mouse Monoclonal Antibody (DyLight® 650) [clone: 6F5]
Lieferant: Novus Biologicals
NB600-444C
NOVUNB600-444CEA 655 EUR
NOVUNB600-444C
Anti-Ethenocytidine Mouse Monoclonal Antibody (DyLight® 650) [clone: 6F5]
Antikörper
Type: Primary
Antigen: Ethenocytidine
Clonality: monoclonal
Clone: 6F5
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG2 Lambda
Reactivity: Pan