Anti-AU5 Goat Polyclonal Antibody

Lieferant: Novus Biologicals
NB600-461
NOVUNB600-461EA 583 EUR
NOVUNB600-461
Anti-AU5 Goat Polyclonal Antibody
Antikörper
Goat Polyclonal AU5 Epitope Tag Antibody. Tested Applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation. Tested Reactivity: Non-species specific.

Type: Primary
Antigen: AU5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: TDFYLK
Host: Goat
Isotype:
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR