Anti-PECAM1 Mouse Monoclonal Antibody [clone: JC/70A]
Lieferant: Novus Biologicals
NB600-562
NOVUNB600-562EA 639 EUR
NOVUNB600-562
Anti-PECAM1 Mouse Monoclonal Antibody [clone: JC/70A]
Antikörper
Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: monoclonal
Clone: JC/70A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen