Anti-GLI1 Rabbit Polyclonal Antibody (DyLight® 650)

Lieferant: Novus Biologicals
NB600-600C
NOVUNB600-600CEA 792 EUR
NOVUNB600-600C
Anti-GLI1 Rabbit Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Primary
Antigen: GLI1 (GLI family zinc finger 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR