Anti-IRF5 Mouse Monoclonal Antibody
Lieferant: Novus Biologicals
NBP1-04307
NOVUNBP1-04307EA 612 EUR
NOVUNBP1-04307
Anti-IRF5 Mouse Monoclonal Antibody
Antikörper
Mouse Monoclonal IRF5 Antibody (10T1). Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation. Tested Reactivity: Human, Mouse.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:50
Type: Primary
Antigen: IRF5
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human